siRNA Details
Gene Symbol: PRPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006445.3 | NP_006436.3 | 43 | 6997 | ||
GenBank mRNA | AB007510.1 | - | 43 | 6924 | ||
AF092565.1 | - | 43 | 6921 | |||
AK058034.1 | - | 11 | 1600 | |||
AK222905.1 | - | 7 | 1073 | |||
AK296344.1 | - | 19 | 3093 | |||
BC000579.2 | - | 9 | 1090 | |||
BC064370.1 | - | 43 | 6962 | 5587081 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
