siRNA Details
Gene Symbol: GFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308164.1 | NP_001295093.1 | 9 | 1214 | ||
NM_001308166.1 | NP_001295095.1 | 8 | 1157 | |||
NM_024996.5 | NP_079272.4 | 8 | 1157 | |||
XM_006713795.1 | XP_006713858.1 | 7 | 1071 | |||
GenBank mRNA | AF309777.1 | - | 8 | 1157 | ||
AF367998.1 | - | 8 | 1157 | |||
AK000780.1 | - | 8 | 1157 | |||
AK022724.1 | - | 3 | 259 | |||
AK092293.1 | - | 7 | 763 | |||
AK315031.1 | - | 8 | 1096 | |||
BC049210.1 | - | 8 | 1121 | 5574223 | ||
KU178816.1 | - | 8 | 1050 | |||
KU178817.1 | - | 7 | 929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
