siRNA Details
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004052.3 | NP_004043.3 | 2 | 273 | ||
GenBank mRNA | AF002697.1 | - | 2 | 192 | ||
AK222626.1 | - | 2 | 183 | |||
AK295136.1 | - | 2 | 183 | |||
BC009342.2 | - | 2 | 168 | |||
BC021989.1 | - | 2 | 187 | 4066029 | ||
BC067818.1 | - | 2 | 216 | |||
BC080643.1 | - | 2 | 168 | |||
HY049371.1 | - | 2 | 274 | |||
U15174.1 | - | 2 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
