siRNA Details
Gene Symbol: FAM71F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282788.1 | NP_001269717.1 | 7 | 1478 | ||
NM_001282789.1 | NP_001269718.1 | 8 | 1425 | |||
NM_032599.3 | NP_115988.1 | 7 | 1484 | |||
NR_104242.1 | 8 | 1782 | ||||
NR_104243.1 | 9 | 1671 | ||||
GenBank mRNA | AF367470.1 | - | 7 | 1437 | ||
AK093161.1 | - | 8 | 1422 | |||
BC037248.1 | - | 7 | 1417 | 5165912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
