siRNA Details
Gene Symbol: RTKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145307.3 | NP_660350.2 | 8, 9 | 1149 | ||
XM_011539456.2 | XP_011537758.1 | 8, 9 | 1165 | |||
XM_011539457.2 | XP_011537759.1 | 8, 9 | 1165 | |||
XM_011539460.2 | XP_011537762.1 | 8, 9 | 1165 | |||
XM_017015842.1 | XP_016871331.1 | 7, 8 | 1067 | |||
XM_017015843.1 | XP_016871332.1 | 8, 9 | 1165 | |||
XM_017015844.1 | XP_016871333.1 | 8, 9 | 1165 | |||
GenBank mRNA | AK096671.1 | - | 3, 4 | 264 | ||
AY150309.1 | - | 8, 9 | 993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
