siRNA Details
Gene Symbol: PGAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170543.1 | NP_001164014.1 | 2 | 399 | ||
NM_001170544.1 | NP_001164015.1 | 2 | 399 | |||
NM_138575.3 | NP_612642.2 | 2 | 399 | |||
GenBank mRNA | AK097688.1 | - | 2 | 746 | ||
BC008196.1 | - | 2 | 356 | 3048106 | ||
BC047906.2 | - | 1 | 54 | |||
DR155760.1 | - | 2 | 367 | |||
EU249757.1 | - | 2 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
