siRNA Details
Gene Symbol: SOHLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282147.1 | NP_001269076.1 | 6 | 715 | ||
NM_017826.2 | NP_060296.2 | 6 | 715 | |||
GenBank mRNA | BC025383.2 | - | 6 | 688 | 4829809 |
Gene Symbol: CCDC169-SOHLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198910.1 | NP_001185839.1 | 11 | 1172 | ||
GenBank mRNA | AK301863.1 | - | 11 | 1159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
