siRNA Details
Gene Symbol: OR13D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004484.1 | NP_001004484.1 | 1 | 938 | ||
GenBank mRNA | BC140916.1 | - | 1 | 981 | 9056728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
