siRNA Details
Gene Symbol: ATAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031722.3 | NP_001026892.1 | 7 | 844 | ||
NM_001190724.2 | NP_001177653.1 | 7 | 844 | |||
NM_001254952.2 | NP_001241881.1 | 7 | 606 | |||
NM_001318762.1 | NP_001305691.1 | 7 | 606 | |||
NM_001318763.1 | NP_001305692.1 | 7 | 606 | |||
NM_024909.3 | NP_079185.2 | 7 | 606 | |||
NR_033823.2 | 7 | 606 | ||||
XM_005249415.2 | XP_005249472.1 | 7 | 807 | |||
XM_017011316.1 | XP_016866805.1 | 7 | 807 | |||
GenBank mRNA | AB075512.1 | - | 7 | 562 | ||
AK023220.1 | - | 7 | 572 | |||
AK225107.1 | - | 7 | 570 | |||
AK297661.1 | - | 7 | 727 | |||
AK298743.1 | - | 7 | 572 | |||
AL535697.3 | - | 7 | 580 | |||
AL833858.1 | - | 7 | 558 | |||
BC006105.1 | - | 7 | 617 | 2820493 | ||
BC025755.1 | - | 7 | 516 | |||
BC047303.1 | - | 7 | 844 | 4914615 | ||
BX442806.2 | - | 7 | 553 | |||
CX786272.1 | - | 7 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
