siRNA Details
Gene Symbol: ZNF771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142305.1 | NP_001135777.1 | 3 | 994 | ||
NM_016643.3 | NP_057727.2 | 3 | 1007 | |||
GenBank mRNA | AF242768.1 | - | 2 | 636 | ||
AK313930.1 | - | 3 | 1114 | |||
BC011870.2 | - | 2 | 756 | 2960099 | ||
BC026192.2 | - | 3 | 793 | 4801104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
