siRNA Details
Gene Symbol: METRNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004431.2 | NP_001004431.1 | 2, 3 | 679 | ||
XM_017024523.1 | XP_016880012.1 | 2, 3 | 1028 | |||
GenBank mRNA | AK093748.1 | - | 1, 2 | 1250 | ||
BC050568.1 | - | 1, 2 | 409 | |||
BC082252.1 | - | 2, 3 | 679 | 6301543 | ||
BC118558.1 | - | 2, 3 | 166 | |||
BC118638.1 | - | 2, 3 | 166 | |||
DR760693.1 | - | 1, 2 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
