siRNA Details
Gene Symbol: WBSCR28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182504.3 | NP_872310.2 | 1 | 95 | ||
GenBank mRNA | AY372053.1 | - | 1 | 100 | ||
AY372054.1 | - | 1 | 22 | |||
BC030643.2 | - | 1 | 95 | 4824662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
