siRNA Details
Gene Symbol: WBSCR28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182504.3 | NP_872310.2 | 3 | 555 | ||
XM_011515785.2 | XP_011514087.1 | 3 | 489 | |||
XM_017011741.1 | XP_016867230.1 | 3 | 677 | |||
GenBank mRNA | AY372053.1 | - | 3 | 560 | ||
AY372054.1 | - | 4 | 606 | |||
BC030643.2 | - | 3 | 555 | 4824662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
