siRNA Details
Gene Symbol: ZNF596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042415.2 | NP_001035880.1 | 6 | 637 | ||
NM_001042416.2 | NP_001035881.1 | 6 | 770 | |||
NM_001287254.1 | NP_001274183.1 | 6 | 628 | |||
NM_001287255.1 | NP_001274184.1 | 6 | 603 | |||
NM_001287256.1 | NP_001274185.1 | 6 | 599 | |||
NM_001287399.1 | NP_001274328.1 | 6 | 686 | |||
NM_173539.3 | NP_775810.2 | 6 | 688 | |||
XM_011534744.1 | XP_011533046.1 | 6 | 610 | |||
XM_017013166.1 | XP_016868655.1 | 5 | 2327 | |||
GenBank mRNA | AK313454.1 | - | 6 | 511 | ||
BC026190.1 | - | 6 | 511 | 4837032 | ||
BG771504.1 | - | 6 | 512 | |||
BM478564.1 | - | 6 | 499 | |||
DB047692.1 | - | 6 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
