siRNA Details
Gene Symbol: CCDC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287181.1 | NP_001274110.1 | 8 | 953 | ||
NM_025055.4 | NP_079331.3 | 18 | 2556 | |||
NM_182791.3 | NP_877592.2 | 9 | 1055 | |||
XM_005254692.1 | XP_005254749.1 | 19 | 2658 | |||
XM_011522085.2 | XP_011520387.1 | 20 | 3503 | |||
XM_011522086.1 | XP_011520388.1 | 19 | 2351 | |||
XM_011522087.2 | XP_011520389.1 | 20 | 2489 | |||
XM_011522088.1 | XP_011520390.1 | 19 | 2359 | |||
XM_011522089.2 | XP_011520391.1 | 13 | 1649 | |||
XM_011522090.2 | XP_011520392.2 | 9 | 1353 | |||
XM_017022623.1 | XP_016878112.1 | 20 | 3504 | |||
XM_017022624.1 | XP_016878113.1 | 19 | 3401 | |||
XM_017022625.1 | XP_016878114.1 | 20 | 3503 | |||
XM_017022626.1 | XP_016878115.1 | 19 | 3401 | |||
XM_017022627.1 | XP_016878116.1 | 20 | 3503 | |||
XM_017022628.1 | XP_016878117.1 | 19 | 3401 | |||
XM_017022629.1 | XP_016878118.1 | 22 | 3244 | |||
XM_017022632.1 | XP_016878121.1 | 9 | 1188 | |||
GenBank mRNA | AK026821.1 | - | 8 | 935 | ||
AK057417.1 | - | 18 | 2442 | |||
AK290709.1 | - | 8 | 953 | |||
BC031684.1 | - | 16 | 1868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
