siRNA Details
Gene Symbol: HSPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002154.3 | NP_002145.3 | 19 | 2697 | ||
GenBank mRNA | AB023420.1 | - | 19 | 2694 | ||
AK294859.1 | - | 19 | 2612 | |||
AK295847.1 | - | 16 | 2082 | |||
AK297762.1 | - | 2 | 228 | |||
AK299710.1 | - | 8 | 973 | |||
AK300050.1 | - | 15 | 1970 | |||
AK300770.1 | - | 18 | 2365 | |||
AK303382.1 | - | 13 | 1617 | |||
AK304578.1 | - | 15 | 1903 | |||
AK311316.1 | - | 7 | 848 | |||
AK315862.1 | - | 5 | 563 | |||
AK315871.1 | - | 4 | 455 | |||
AK315875.1 | - | 3 | 378 | |||
AK315887.1 | - | 7 | 852 | |||
BC002526.2 | - | 5 | 540 | |||
BC110861.1 | - | 19 | 2557 | 6169048 | ||
BC126122.1 | - | 19 | 2658 | 8991838 | ||
BC126124.1 | - | 19 | 2658 | 8991840 | ||
BQ431980.1 | - | 6 | 633 | |||
BT007375.1 | - | 5 | 340 | |||
BU171424.1 | - | 1 | 53 | |||
BU686725.1 | - | 2 | 160 | |||
CF145856.1 | - | 2 | 221 | |||
FJ224293.1 | - | 19 | 2694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
