siRNA Details
Gene Symbol: TM6SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144903.1 | NP_001138375.1 | 9 | 1041 | ||
NM_023003.3 | NP_075379.2 | 10 | 1134 | |||
XM_011521680.2 | XP_011519982.1 | 11 | 1183 | |||
XM_017022323.1 | XP_016877812.1 | 10 | 1186 | |||
XM_017022324.1 | XP_016877813.1 | 9 | 1070 | |||
XM_017022326.1 | XP_016877815.1 | 10 | 1048 | |||
XM_017022327.1 | XP_016877816.1 | 10 | 1240 | |||
XM_017022328.1 | XP_016877817.1 | 7 | 809 | |||
GenBank mRNA | AK223246.1 | - | 10 | 1079 | ||
AK292100.1 | - | 10 | 1059 | |||
AK295568.1 | - | 10 | 1023 | |||
BC032007.1 | - | 6 | 645 | |||
BC063487.1 | - | 7 | 814 | 5240735 | ||
DQ021909.1 | - | 9 | 932 |

More Information
Other Species Targeted

Gene Ontology Categories:
