siRNA Details
Gene Symbol: TM6SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144903.1 | NP_001138375.1 | 9 | 1135 | ||
NM_023003.3 | NP_075379.2 | 10 | 1228 | |||
XM_011521680.2 | XP_011519982.1 | 11 | 1277 | |||
XM_017022323.1 | XP_016877812.1 | 10 | 1280 | |||
XM_017022324.1 | XP_016877813.1 | 9 | 1164 | |||
XM_017022326.1 | XP_016877815.1 | 10 | 1142 | |||
XM_017022327.1 | XP_016877816.1 | 10 | 1334 | |||
XM_017022328.1 | XP_016877817.1 | 7 | 903 | |||
GenBank mRNA | AF255922.1 | - | 10 | 1176 | ||
AK223246.1 | - | 10 | 1173 | |||
AK292100.1 | - | 10 | 1153 | |||
AK295568.1 | - | 10 | 1117 | |||
BC032007.1 | - | 6 | 739 | |||
BC063487.1 | - | 7 | 908 | 5240735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
