siRNA Details
Gene Symbol: MYADML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003143.2 | 1 | 1109 | |||
GenBank mRNA | AK057470.1 | - | 1 | 1096 | ||
BC031088.1 | - | 1 | 1099 | |||
BC038980.1 | - | 1 | 1070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
