siRNA Details
Gene Symbol: TPI1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002187.3 | 1 | 915 | |||
GenBank mRNA | AK094894.1 | - | 1 | 917 | ||
BC053624.1 | - | 1 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
