siRNA Details
Gene Symbol: DNAJC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032317.2 | NP_115693.2 | 1 | 621 | ||
GenBank mRNA | AF412025.1 | - | 1 | 612 | ||
AK058113.1 | - | 1 | 601 | |||
AK094305.1 | - | 1 | 621 | |||
BC005056.1 | - | 1 | 612 | 2823228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
