siRNA Details
Gene Symbol: LRRC75A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113567.2 | NP_001107039.1 | 4 | 2542 | ||
NM_207387.3 | NP_997270.2 | 3 | 2426 | |||
XM_011523845.2 | XP_011522147.1 | 5 | 2615 | |||
XM_017024619.1 | XP_016880108.1 | 4 | 2311 | |||
XM_017024620.1 | XP_016880109.1 | 5 | 2474 | |||
GenBank mRNA | AK093015.1 | - | 3 | 2085 | ||
AL833245.1 | - | 3 | 2452 | |||
AL833260.1 | - | 3 | 2452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
