siRNA Details
Gene Symbol: MLLT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005937.3 | NP_005928.2 | 14 | 2056 | ||
XM_006721911.2 | XP_006721974.1 | 14 | 1978 | |||
XM_017024648.1 | XP_016880137.1 | 9 | 1581 | |||
GenBank mRNA | AB209633.1 | - | 14 | 2031 | ||
AK027133.1 | - | 3 | 174 | |||
AL133659.1 | - | 2 | 1035 | |||
BC007237.1 | - | 6 | 1007 | |||
BC007859.2 | - | 4 | 407 | |||
U07932.1 | - | 14 | 2043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
