siRNA Details
Gene Symbol: DCAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163484.1 | NP_001156956.1 | 9 | 1100 | ||
NM_025230.4 | NP_079506.3 | 9 | 1550 | |||
NM_181357.2 | NP_852002.1 | 9 | 902 | |||
NR_028099.1 | 8 | 1462 | ||||
NR_028100.1 | 8 | 1077 | ||||
GenBank mRNA | AB209073.1 | - | 8 | 1448 | ||
AF130070.1 | - | 8 | 810 | |||
AF267858.1 | - | 9 | 1213 | |||
AK027223.1 | - | 2 | 79 | |||
AK057636.1 | - | 9 | 861 | |||
AK074296.1 | - | 9 | 1515 | |||
AK091857.1 | - | 1 | 642 | |||
AK292797.1 | - | 9 | 982 | |||
AK296742.1 | - | 8 | 1034 | |||
AK307849.1 | - | 6 | 682 | |||
AL833831.1 | - | 2 | 67 | |||
BC008858.2 | - | 4 | 328 | |||
BC067132.1 | - | 9 | 973 | 6170335 | ||
BX161405.1 | - | 8 | 797 | |||
BX247970.1 | - | 7 | 976 | |||
BX648224.1 | - | 9 | 1032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
