siRNA Details
Gene Symbol: DCAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163484.1 | NP_001156956.1 | 15 | 2275 | ||
NM_025230.4 | NP_079506.3 | 15 | 2725 | |||
NM_181357.2 | NP_852002.1 | 15 | 2077 | |||
NR_028099.1 | 14 | 2637 | ||||
NR_028100.1 | 14 | 2252 | ||||
GenBank mRNA | AB209073.1 | - | 14 | 2623 | ||
AF130070.1 | - | 14 | 1985 | |||
AF267858.1 | - | 15 | 2395 | |||
AK027223.1 | - | 8 | 1254 | |||
AK057636.1 | - | 15 | 2036 | |||
AK074296.1 | - | 15 | 2690 | |||
AK091857.1 | - | 5 | 2259 | |||
AK094478.1 | - | 9 | 1172 | |||
AK296742.1 | - | 14 | 2209 | |||
AL833831.1 | - | 8 | 1242 | |||
BC008858.2 | - | 10 | 1506 | |||
BC067132.1 | - | 15 | 2148 | 6170335 | ||
BX161405.1 | - | 14 | 1972 | |||
BX247970.1 | - | 13 | 2151 | |||
BX648224.1 | - | 15 | 2207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
