siRNA Details
Gene Symbol: DCAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163484.1 | NP_001156956.1 | 3, 4 | 563 | ||
NM_025230.4 | NP_079506.3 | 3, 4 | 1013 | |||
NM_181357.2 | NP_852002.1 | 3, 4 | 365 | |||
NR_028099.1 | 3, 4 | 990 | ||||
NR_028100.1 | 2, 3 | 540 | ||||
GenBank mRNA | AB209073.1 | - | 3, 4 | 976 | ||
AF130070.1 | - | 2, 3 | 273 | |||
AF267858.1 | - | 3, 4 | 676 | |||
AK057636.1 | - | 3, 4 | 324 | |||
AK074296.1 | - | 3, 4 | 978 | |||
AK292797.1 | - | 3, 4 | 445 | |||
AK296742.1 | - | 2, 3 | 497 | |||
BC067132.1 | - | 3, 4 | 436 | 6170335 | ||
BX161405.1 | - | 2, 3 | 260 | |||
BX247970.1 | - | 2, 3 | 504 | |||
BX648224.1 | - | 3, 4 | 495 | |||
DB212727.1 | - | 3, 4 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
