siRNA Details
Gene Symbol: EID2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152361.2 | NP_689574.1 | 1 | 82 | ||
GenBank mRNA | AK096263.1 | - | 1 | 69 | ||
BC009725.1 | - | 1 | 70 | 3897193 | ||
BC040157.1 | - | 1 | 26 | |||
BC051333.1 | - | 1 | 43 | |||
DA382981.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
