siRNA Details
Gene Symbol: TNFAIP8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311175.1 | NP_001298104.1 | 2 | 672 | ||
NM_207381.3 | NP_997264.2 | 3 | 926 | |||
XM_011521545.2 | XP_011519847.1 | 2 | 679 | |||
XM_017022168.1 | XP_016877657.1 | 2 | 1025 | |||
XM_017022169.1 | XP_016877658.1 | 2 | 600 | |||
GenBank mRNA | AK123281.1 | - | 2 | 656 | ||
AY517501.1 | - | 3 | 926 | |||
BC127701.1 | - | 2 | 586 | 40127272 | ||
BC127702.1 | - | 2 | 586 | 40127273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
