siRNA Details
Gene Symbol: TNFAIP8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311175.1 | NP_001298104.1 | 2 | 456 | ||
NM_207381.3 | NP_997264.2 | 3 | 710 | |||
XM_011521545.2 | XP_011519847.1 | 2 | 463 | |||
XM_017022168.1 | XP_016877657.1 | 2 | 809 | |||
XM_017022169.1 | XP_016877658.1 | 2 | 384 | |||
GenBank mRNA | AK123281.1 | - | 2 | 440 | ||
AY517501.1 | - | 3 | 710 | |||
BC127701.1 | - | 2 | 370 | 40127272 | ||
BC127702.1 | - | 2 | 370 | 40127273 |

More Information
Other Species Targeted

Gene Ontology Categories:
