siRNA Details
Gene Symbol: GMCL1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003281.1 | 1 | 1665 | |||
GenBank mRNA | AF198534.1 | - | 1 | 1115 | ||
AK315701.1 | - | 1 | 1635 | |||
BC024184.2 | - | 1 | 1651 | |||
BC024185.1 | - | 1 | 1659 | |||
BC033886.2 | - | 1 | 1665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
