siRNA Details
Gene Symbol: LINC00346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027701.1 | 1 | 604 | |||
GenBank mRNA | AJ295981.1 | - | 1 | 644 | ||
AK307956.1 | - | 1 | 519 | |||
AL831965.2 | - | 1 | 563 | |||
BC093904.2 | - | 1 | 111 | 7939749 | ||
BC104998.1 | - | 1 | 111 | 8144001 | ||
BX647632.1 | - | 1 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
