siRNA Details
Gene Symbol: C19orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199341.3 | NP_955373.3 | 2 | 282 | ||
XM_005259449.3 | XP_005259506.1 | 2 | 720 | |||
XM_006723204.3 | XP_006723267.1 | 2 | 417 | |||
XM_006723205.2 | XP_006723268.1 | 2 | 308 | |||
GenBank mRNA | BC043386.1 | - | 2 | 282 | 5492230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
