siRNA Details
Gene Symbol: SLC35F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300845.1 | NP_001287774.1 | 5 | 1147 | ||
NM_173508.3 | NP_775779.1 | 6 | 1432 | |||
GenBank mRNA | AK095031.1 | - | 6 | 1213 | ||
BC037878.1 | - | 5 | 1091 | 5273868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
