siRNA Details
Gene Symbol: PPP4R3CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002784.2 | 1 | 1641 | |||
GenBank mRNA | AK057429.1 | - | 1 | 716 | ||
AK131475.1 | - | 1 | 1641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
