siRNA Details
Gene Symbol: ACSM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105069.1 | NP_001098539.1 | 14 | 2051 | ||
NM_182617.3 | NP_872423.3 | 15 | 2086 | |||
XM_011545824.2 | XP_011544126.1 | 13 | 1872 | |||
GenBank mRNA | AK129944.1 | - | 14 | 1990 | ||
BC026010.1 | - | 1 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
