siRNA Details
Gene Symbol: ACSM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105069.1 | NP_001098539.1 | 9, 10 | 1360 | ||
NM_182617.3 | NP_872423.3 | 10, 11 | 1395 | |||
XM_005255304.3 | XP_005255361.1 | 9, 10 | 1299 | |||
XM_011545824.2 | XP_011544126.1 | 8, 9 | 1181 | |||
GenBank mRNA | AB073604.1 | - | 10, 11 | 1326 | ||
AK129944.1 | - | 9, 10 | 1299 | |||
AK289416.1 | - | 9, 10 | 1299 | |||
AK309601.1 | - | 9, 10 | 1360 | |||
AY160217.1 | - | 8, 9 | 1185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
