siRNA Details
Gene Symbol: CCDC190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178550.4 | NP_848645.3 | 3 | 389 | ||
XM_005245125.3 | XP_005245182.1 | 3 | 3007 | |||
XM_006711289.3 | XP_006711352.1 | 3 | 489 | |||
XM_011509477.2 | XP_011507779.1 | 3 | 3010 | |||
GenBank mRNA | AK123573.1 | - | 3 | 386 | ||
AK309620.1 | - | 1 | 52 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
