siRNA Details
Gene Symbol: FOXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023067.3 | NP_075555.1 | 1 | 1028 | ||
GenBank mRNA | AF301906.1 | - | 1 | 847 | ||
BC019896.1 | - | 1 | 775 | |||
BC062549.1 | - | 1 | 1028 | 6572303 | ||
DQ089670.1 | - | 1 | 847 | |||
DQ089671.1 | - | 1 | 847 | |||
DQ089672.1 | - | 1 | 847 | |||
FJ598614.1 | - | 1 | 847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
