siRNA Details
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033184.1 | 3 | 414 | |||
NR_102302.1 | 4 | 507 | ||||
GenBank mRNA | AK307248.1 | - | 4 | 483 | ||
BC042500.1 | - | 3 | 390 | 4823179 | ||
DA213851.1 | - | 3 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
