siRNA Details
Gene Symbol: DLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317172.1 | NP_001304101.1 | 4 | 533 | ||
NM_003836.6 | NP_003827.3 | 4 | 533 | |||
GenBank mRNA | AK289384.1 | - | 4 | 503 | ||
AK297918.1 | - | 3 | 429 | |||
AK313258.1 | - | 4 | 503 | |||
BC007741.2 | - | 4 | 482 | 4128269 | ||
BC013197.1 | - | 4 | 449 | 4347187 | ||
BC014015.2 | - | 4 | 478 | 4130556 | ||
DQ017081.1 | - | 4 | 329 | |||
DQ126100.1 | - | 4 | 329 | |||
DQ138965.1 | - | 4 | 329 | |||
DQ309460.1 | - | 1 | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
