siRNA Details
Gene Symbol: DND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194249.2 | NP_919225.1 | 3 | 358 | ||
GenBank mRNA | AY321065.1 | - | 3 | 314 | ||
BC033496.1 | - | 3 | 358 | 5172595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
