siRNA Details
Gene Symbol: SLC25A45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077241.2 | NP_001070709.2 | 4, 5 | 499 | ||
NM_001278250.2 | NP_001265179.2 | 3, 4 | 710 | |||
NM_001278251.2 | NP_001265180.2 | 8, 9 | 1356 | |||
NM_182556.3 | NP_872362.3 | 5, 6 | 554 | |||
XM_006718509.3 | XP_006718572.1 | 4, 5 | 1164 | |||
XM_006718510.3 | XP_006718573.1 | 6, 7 | 692 | |||
XM_011544943.2 | XP_011543245.1 | 5, 6 | 1263 | |||
XM_011544944.2 | XP_011543246.1 | 4, 5 | 593 | |||
XM_011544947.2 | XP_011543249.1 | 4, 5 | 782 | |||
XM_011544949.2 | XP_011543251.1 | 6, 7 | 611 | |||
XM_017017562.1 | XP_016873051.1 | 4, 5 | 782 | |||
XM_017017563.1 | XP_016873052.1 | 5, 6 | 2029 | |||
XM_017017564.1 | XP_016873053.1 | 5, 6 | 2028 | |||
XM_017017565.1 | XP_016873054.1 | 5, 6 | 1293 | |||
XM_017017566.1 | XP_016873055.1 | 5, 6 | 648 | |||
XM_017017567.1 | XP_016873056.1 | 6, 7 | 654 | |||
XM_017017568.1 | XP_016873057.1 | 5, 6 | 477 | |||
XM_017017569.1 | XP_016873058.1 | 7, 8 | 1356 | |||
GenBank mRNA | AK090434.1 | - | 1 | 2611 | ||
AK091190.1 | - | 8, 9 | 1356 | |||
AK124022.1 | - | 1 | 957 | |||
BC036869.1 | - | 4, 5 | 462 | 5228549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
