siRNA Details
Gene Symbol: SLC25A45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006718509.3 | XP_006718572.1 | 6 | 1480 | ||
XM_006718510.3 | XP_006718573.1 | 8 | 1008 | |||
XM_011544943.2 | XP_011543245.1 | 7 | 1579 | |||
XM_011544944.2 | XP_011543246.1 | 6 | 909 | |||
XM_011544947.2 | XP_011543249.1 | 6 | 1098 | |||
XM_011544949.2 | XP_011543251.1 | 8 | 927 | |||
XM_017017562.1 | XP_016873051.1 | 6 | 1098 | |||
XM_017017563.1 | XP_016873052.1 | 7 | 2345 | |||
XM_017017564.1 | XP_016873053.1 | 7 | 2344 | |||
XM_017017565.1 | XP_016873054.1 | 7 | 1609 | |||
XM_017017566.1 | XP_016873055.1 | 7 | 964 | |||
XM_017017567.1 | XP_016873056.1 | 8 | 970 | |||
XM_017017568.1 | XP_016873057.1 | 7 | 793 | |||
XM_017017569.1 | XP_016873058.1 | 9 | 1672 | |||
XM_017017570.1 | XP_016873059.1 | 5 | 1295 | |||
XM_017017571.1 | XP_016873060.1 | 5 | 665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
