siRNA Details
Gene Symbol: IQCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203424.1 | NP_982248.1 | 3 | 161 | ||
GenBank mRNA | AK128883.1 | - | 3 | 202 | ||
BC040047.1 | - | 3 | 152 | 5742210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
