siRNA Details
Gene Symbol: LEMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199050.1 | NP_001185979.1 | 4 | 395 | ||
NM_001199051.1 | NP_001185980.1 | 3 | 272 | |||
XM_011510160.2 | XP_011508462.1 | 4 | 399 | |||
XM_011510161.2 | XP_011508463.1 | 4 | 387 | |||
XM_011510162.2 | XP_011508464.1 | 4 | 397 | |||
XM_011510163.2 | XP_011508465.1 | 3 | 279 | |||
XM_011510164.2 | XP_011508466.1 | 4 | 419 | |||
XM_011510165.2 | XP_011508467.1 | 4 | 415 | |||
GenBank mRNA | AB096677.1 | - | 4 | 362 | ||
AB096679.1 | - | 3 | 239 | |||
AK302392.1 | - | 4 | 397 | |||
BC036636.2 | - | 4 | 365 | 5296437 | ||
DC399934.1 | - | 4 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
