siRNA Details
Gene Symbol: ZDHHC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185024.1 | NP_001171953.1 | 7 | 960 | ||
NM_013373.3 | NP_037505.1 | 7 | 960 | |||
XM_006724239.2 | XP_006724302.1 | 7 | 948 | |||
GenBank mRNA | AB033118.1 | - | 3 | 344 | ||
AK131238.1 | - | 7 | 811 | |||
AY894890.1 | - | 7 | 960 | |||
BC053544.1 | - | 7 | 960 | 5730191 | ||
CR456357.1 | - | 6 | 717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
