siRNA Details
Gene Symbol: ZDHHC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013373.3 | NP_037505.1 | 11 | 2391 | ||
GenBank mRNA | AB033118.1 | - | 7 | 1775 | ||
AY894890.1 | - | 11 | 2391 | |||
BC053544.1 | - | 11 | 2391 | 5730191 | ||
CR456357.1 | - | 10 | 2148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
