siRNA Details
Gene Symbol: LRRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033402.4 | NP_208325.3 | 9 | 1596 | ||
XM_005251318.3 | XP_005251375.1 | 8 | 1385 | |||
XM_005251319.4 | XP_005251376.1 | 7 | 1079 | |||
XM_005251320.3 | XP_005251377.1 | 6 | 871 | |||
XM_005251322.3 | XP_005251379.1 | 5 | 685 | |||
XM_017013920.1 | XP_016869409.1 | 10 | 1544 | |||
XM_017013921.1 | XP_016869410.1 | 10 | 1544 | |||
XM_017013922.1 | XP_016869411.1 | 10 | 1544 | |||
XM_017013923.1 | XP_016869412.1 | 9 | 1371 | |||
XM_017013924.1 | XP_016869413.1 | 10 | 1544 | |||
XM_017013925.1 | XP_016869414.1 | 7 | 1090 | |||
XM_017013926.1 | XP_016869415.1 | 7 | 1073 | |||
XM_017013927.1 | XP_016869416.1 | 8 | 1101 | |||
XM_017013928.1 | XP_016869417.1 | 5 | 553 | |||
GenBank mRNA | AB051551.1 | - | 8 | 2217 | ||
AK056185.1 | - | 8 | 1559 | |||
AK300874.1 | - | 8 | 1240 | |||
AK302648.1 | - | 9 | 1446 | |||
AK307350.1 | - | 5 | 660 | |||
AK307443.1 | - | 6 | 852 | |||
AK307636.1 | - | 8 | 1230 | |||
BC030701.2 | - | 9 | 1469 | 4837403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
