siRNA Details
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 7 | 805 | ||
XM_011538083.1 | XP_011536385.1 | 7 | 669 | |||
XM_017019091.1 | XP_016874580.1 | 7 | 645 | |||
GenBank mRNA | AB023163.1 | - | 7 | 749 | ||
AB024494.1 | - | 7 | 1295 | |||
AB097013.1 | - | 7 | 625 | |||
AB097035.1 | - | 7 | 625 | |||
AK292866.1 | - | 7 | 621 | |||
AK299089.1 | - | 6 | 517 | |||
BC030990.1 | - | 6 | 672 | |||
BC050324.1 | - | 7 | 764 | 5266097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
