siRNA Details
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 6 | 725 | ||
XM_005268750.4 | XP_005268807.1 | 6 | 565 | |||
XM_011538083.1 | XP_011536385.1 | 6 | 589 | |||
XM_017019091.1 | XP_016874580.1 | 6 | 565 | |||
XM_017019092.1 | XP_016874581.1 | 5 | 469 | |||
XM_017019093.1 | XP_016874582.1 | 5 | 418 | |||
XM_017019094.1 | XP_016874583.1 | 5 | 446 | |||
XM_017019095.1 | XP_016874584.1 | 2 | 158 | |||
GenBank mRNA | AB023163.1 | - | 6 | 669 | ||
AB024494.1 | - | 6 | 1215 | |||
AB097013.1 | - | 6 | 545 | |||
AB097035.1 | - | 6 | 545 | |||
AK292866.1 | - | 6 | 541 | |||
AK299089.1 | - | 5 | 437 | |||
BC030990.1 | - | 5 | 592 | |||
BC050324.1 | - | 6 | 684 | 5266097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
